EMAIL THIS PAGE TO A FRIEND
F8502 Sigma-Aldrich

N-Formyl-Met-Ala

≥95% (HPLC)

  • CAS Number 15183-28-5

  • Empirical Formula (Hill Notation) C9H16N2O4S

  • Molecular Weight 248.30

  •  MDL number MFCD00057572

  •  PubChem Substance ID 24894964

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
assay   ≥95% (HPLC)
storage temp.   −20°C
SMILES string   CSCCC(NC=O)C(=O)NC(C)C(O)=O
InChI   1S/C9H16N2O4S/c1-6(9(14)15)11-8(13)7(10-5-12)3-4-16-2/h5-7H,3-4H2,1-2H3,(H,10,12)(H,11,13)(H,14,15)
InChI key   GBWVAAKKEIOROG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?