EMAIL THIS PAGE TO A FRIEND
F9376 Sigma-Aldrich

Nε-Formyl-L-lysine

  • CAS Number 1190-48-3

  • Empirical Formula (Hill Notation) C7H14N2O3

  • Molecular Weight 174.20

  •  MDL number MFCD00037362

  •  PubChem Substance ID 24894995

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   KLPJXDPPMSJWKI-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Nε-Formyl-L-lysine is an analogue of L-lysine used as a reference material in the detection of glycated lysine molecules generated by ribose, lactose and fructose Maillard reactions.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?