EMAIL THIS PAGE TO A FRIEND
F9420 Sigma

Fmoc-Tic-OH

≥95% (TLC)

Synonym: (S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • CAS Number 136030-33-6

  • Empirical Formula (Hill Notation) C25H21NO4

  • Molecular Weight 399.44

  •  Beilstein Registry Number 4827574

  •  MDL number MFCD00144368

Purchase

Properties

InChI Key   LIRBCUNCXDZOOU-QHCPKHFHSA-N
assay   ≥95% (TLC)
storage temp.   −20°C

Description

Linkage

Bridged analog of FMOC-L-phenylalanine

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

47701 Fmoc-Tic-OH, ≥97.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?