• USA Home
  • G0422 - Anti-GPI (211-225) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G0422 Sigma

Anti-GPI (211-225) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-AMF, Anti-GNPI, Anti-Glucose phosphate isomerase, Anti-NLK, Anti-PGI, Anti-PHI, Anti-SA-36

Purchase

Properties

Related Categories Aerobic Glycolysis (the Warburg Effect), Alphabetical Index, Antibodies, Antibodies Against Proteins Involved in Glucose Metabolism, Cancer Metabolism,
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~63 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GPI(2821)
biological source   rabbit
antibody product type   primary antibodies
conjugate   unconjugated
UniProt accession no.   P06744

Description

Immunogen

synthetic peptide corresponding to amino acids 211-225 of human GPI.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-GPI (211-225) antibody is suitable for western blot at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

GPI acts as a glycolytic enzyme as well as neurotrophic factor (Neuroleukin) for spinal and sensory neurons. It is associated with various biological processes such as cytokine activity, mitogenesis, differentiation, oncogenesis, and tumor metastasis. By catalyzing the isomerization between glucose-6-phosphate and fructose-6-phosphate, GPI plays a key role in both glycolysis and gluconeogenesis pathways. In mammals, it functions as a tumor-secreted cytokine and an angiogenic factor (AMF) which helps to stimulate endothelial cell motility. Deficiency in GPI causes an autosomal recessive disorder, which results in nonspherocytic hemolytic anemia.

Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Articles

Aerobic Glycolysis and the Warburg Effect

The Warburg effect is the enhanced conversion of glucose to lactate observed in tumor cells, even in the presence of normal levels of oxygen. Otto Heinrich Warburg demonstrated in 1924 that cancer ce...
Keywords: Aerobic, Cancer, Cell proliferation, Clinical, Glycolysis, PAGE, Phosphorylations, Transcription

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?