EMAIL THIS PAGE TO A FRIEND
G0642 Sigma

Gly-Ser-Phe

  • CAS Number 87494-17-5

  • Empirical Formula (Hill Notation) C14H19N3O5

  • Molecular Weight 309.32

  •  MDL number MFCD00057932

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?