• USA Home
  • G0648 - L-Glutamic acid diallyl ester p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
G0648 Sigma-Aldrich

L-Glutamic acid diallyl ester p-toluenesulfonate salt

Synonym: L-Glutamic acid bis-allyl ester p-toluenesulfonate salt

  • CAS Number 20845-16-3

  • Linear Formula CH2=CHCH2OCOCH2CH2CH(NH2)COOCH2CH=CH2 · C7H8O3S

  • Molecular Weight 399.46

  •  MDL number MFCD00043307

Purchase

Properties

storage temp.   −20°C
SMILES string   Cc1ccc(cc1)S([O-])(=O)=O.[NH3+][C@@H](CCC(=O)OCC=C)C(=O)OCC=C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?