• USA Home
  • G1769 - β-L-Glucose pentaacetate

EMAIL THIS PAGE TO A FRIEND
G1769 Sigma-Aldrich

β-L-Glucose pentaacetate

  • CAS Number 66966-07-2

  • Empirical Formula (Hill Notation) C16H22O11

  • Molecular Weight 390.34

  •  MDL number MFCD00069796

Purchase

Properties

SMILES string   CC(=O)OC[C@@H]1O[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?