EMAIL THIS PAGE TO A FRIEND
G3376 Sigma-Aldrich

Glu-Ala

  • CAS Number 21064-18-6

  • Empirical Formula (Hill Notation) C8H14N2O5

  • Molecular Weight 218.21

  •  MDL number MFCD00037233

Purchase

Properties

storage temp.   −20°C
SMILES string   C[C@H](NC(=O)[C@@H](N)CCC(O)=O)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?