EMAIL THIS PAGE TO A FRIEND
G3501 Sigma

Glu-Gly-Phe

  • CAS Number 42155-93-1

  • Empirical Formula (Hill Notation) C16H21N3O6

  • Molecular Weight 351.35

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?