• USA Home
  • G3515 - Gly-Pro p-nitroanilide p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
G3515

Gly-Pro p-nitroanilide p-toluenesulfonate salt

  • CAS Number 65096-46-0

  • Empirical Formula (Hill Notation) C13H16N4O4 · C7H8O3S

  • Molecular Weight 464.49

  •  MDL number MFCD00039007

Purchase

Properties

storage temp.   −20°C
SMILES string   Cc1ccc(cc1)S(O)(=O)=O.NCC(=O)N2CCC[C@H]2C(=O)Nc3ccc(cc3)[N+]([O-])=O
InChI   1S/C13H16N4O4.C7H8O3S/c14-8-12(18)16-7-1-2-11(16)13(19)15-9-3-5-10(6-4-9)17(20)21;1-6-2-4-7(5-3-6)11(8,9)10/h3-6,11H,1-2,7-8,14H2,(H,15,19);2-5H,1H3,(H,8,9,10)/t11-;/m0./s1
InChI key   XHLUWCXVWCBOPP-MERQFXBCSA-N

Description

Substrates

Substrate for Dipeptidyl Peptidase IV.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?