• USA Home
  • G3535 - [D-Thr6, D-Trp8,9, 15-ol]-Galanin fragment 1-15

EMAIL THIS PAGE TO A FRIEND
G3535 Sigma-Aldrich

[D-Thr6, D-Trp8,9, 15-ol]-Galanin fragment 1-15

≥97% (HPLC)

  • CAS Number 150940-97-9

  • Empirical Formula (Hill Notation) C84H117N21O18

  • Molecular Weight 1708.96

  •  MDL number MFCD01861110

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](C)NC(=O)[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)CN)[C@@H](C)O)[C@H](C)O)C(=O)NCC(=O)N7CCC[C@H]7C(=O)N[C@@H](Cc8c[nH]cn8)C(=O)N[C@@H](C)CO
Gene Information   human ... GAL(51083)

Description

Amino Acid Sequence

Gly-Trp-Thr-Leu-Asn-D-Thr-Ala-D-Trp-D-Trp-Leu-Leu-Gly-Pro-His-Ala-ol

Biochem/physiol Actions

A potent galanin antagonist at sub-micromolar concentrations.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?