• USA Home
  • G3629 - Glycine ethyl ester phosphate salt

EMAIL THIS PAGE TO A FRIEND
G3629 Sigma-Aldrich

Glycine ethyl ester phosphate salt

Synonym: Ethyl glycinate phosphate salt

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?