EMAIL THIS PAGE TO A FRIEND
G4395 Sigma-Aldrich

D-Glucose-UL-14C

≥98% radiochemical purity basis (HPLC), aqueous solution

Purchase

Properties

assay   ≥98% radiochemical purity basis (HPLC)
form   aqueous solution
extent of labeling   2-10 mCi per mmol
mass shift   M+2
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Combi-vial

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Glycolysis Overview

Carbohydrates synthesized during photosynthesis act as the main storage molecules of solar energy. When ingested, complex carbohydrates are enzymatically hydrolyzed to monosaccharides, such as starch...
BioFiles 2007, 2.6, 4.
Keywords: Aldol reaction, Dehydration reaction, Glycolysis, Isomerizations, Metabolism, Metabolites, Metabolomics, Phosphorylations, Photosynthesis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?