• USA Home
  • G5008 - L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt

EMAIL THIS PAGE TO A FRIEND
G5008 Sigma-Aldrich

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt

≥98% (TLC)

Synonym: L-Glutamic acid 5-(3-carboxy-4-nitroanilide) ammonium salt

Purchase

Properties

assay   ≥98% (TLC)
storage temp.   2-8°C
SMILES string   N.N[C@@H](CCC(=O)Nc1ccc(c(c1)C(O)=O)[N+]([O-])=O)C(O)=O
InChI   1S/C12H13N3O7.H3N/c13-8(12(19)20)2-4-10(16)14-6-1-3-9(15(21)22)7(5-6)11(17)18;/h1,3,5,8H,2,4,13H2,(H,14,16)(H,17,18)(H,19,20);1H3/t8-;/m0./s1
InChI key   GFABNNMTRVBLPZ-QRPNPIFTSA-N

Description

Quality

Tested for suitability as a substrate for γ-glutamyltranspeptidase.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

49525 L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt, ≥99.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?