• USA Home
  • G5513 - DL-Glutamic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
G5513 Sigma-Aldrich

DL-Glutamic acid monohydrate

suitable for cell culture, BioReagent

Synonym: (±)-2-Aminoglutaric acid, (±)-2-Aminopentanedioic acid

  • CAS Number 19285-83-7

  • Linear Formula HOOCCH2CH2CH(NH2)COOH · H2O

  • Molecular Weight 165.14

  •  Beilstein/REAXYS Number 4155097

  •  EC Number 210-522-2

  •  MDL number MFCD00150703

  •  PubChem Substance ID 24895215

Purchase

Properties

Related Categories Amino Acids, Amino Acids and Vitamins, Cell Culture, Reagents and Supplements
InChI Key   OZDAOHVKBFBBMZ-UHFFFAOYSA-N
product line   BioReagent
suitability   suitable for cell culture

Description

General description

L-Glutamic acid is a proteinogenic amino acid incorporated into proteins as directed by the genetic code. This product is a mixture of D and L isomers of glutamic acid.

Packaging

1 kg in poly bottle

100 g in poly bottle

Application

Qualified for use in cell culture applications where mixed isomers are indicated.

Biochem/physiol Actions

Excitatory amino acid neurotransmitter that is an agonist at kainate, NMDA, AMPA, and metabotropic glutamatergic receptors.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Request a Serum Sample

Cell Culture Guide
Protocols & Articles

Articles

HPLC Analysis of FMOC-Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of FMOC-Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T (23 °C)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T (23 °C)
Keywords: Chromatography, High performance liquid chromatography

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?