EMAIL THIS PAGE TO A FRIEND
G5667 SAFC

L-Glutamic acid

PharmaGrade, Ajinomoto, EP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture

Synonym: (S)-2-Aminopentanedioic acid, Glu

  • CAS Number 56-86-0

  • Linear Formula HO2CCH2CH2CH(NH2)CO2H

  • Molecular Weight 147.13

  •  Beilstein/REAXYS Number 1723801

  •  EC Number 200-293-7

  •  MDL number MFCD00002634

  •  PubChem Substance ID 24895223

Purchase

Properties

Related Categories Amino Acids, Analytical/Chromatography, Biochemicals and Reagents, Pharmacopoeia, Pharmacopoeia A-Z,
Quality Level   GMP-COMPENDIA
biological source   non-animal source
grade   EP
InChI Key   WHUUTDBJXJRKMK-VKHMYHEASA-N
product line   PharmaGrade
form   crystalline powder
mfr. no.   Ajinomoto
storage condition   room temperature
impurities   endotoxin, heavy metals, residual solvents, tested
mp   205 °C (dec.) (lit.)
solubility   1 M HCl: 100 mg/mL
suitability   suitable for cell culture
  suitable for manufacturing use
Featured Industry   Pharmaceutical (small molecule)
foreign activity   cytotoxicity, tested
Gene Information   human ... CCR2(1231), GRIA2(2891), GRIA4(2893), GRIK1(2897), GRIK2(2898), GRIK3(2899), GRIK5(2901), GRIN2B(2904), GRM2(2912), SLC1A1(6505), SLC1A2(6506)
rat ... Gria1(50592), Grik1(29559), Grik2(54257), Grik4(24406), Grin2a(24409), Grm1(24414), Grm2(24415), Grm3(24416), Grm4(24417), Grm5(24418), Grm6(24419), Grm7(81672), Slc1a2(29482)

Description

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Application

L-Glutamic acid is a non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Biochem/physiol Actions

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Linkage

To request the Enhanced Quality Product Documentation for this product, submit a PharmaGrade Product Information Request Form. Please note that access to the Enhanced Quality Product Documentation for this product requires a confidentiality disclosure agreement.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

Amino Acids Reference Chart

Amino acids are the 21 standard compounds or building blocks that make up peptides and proteins, including DNA. Each amino acid is structured from an amino group and a carboxyl group bound to a tetra...
Keywords: Building blocks, Gene expression, Mass spectrometry

Enhanced Quality Program [VIDEO]

The Enhanced Quality Program is a quality management system that is used to differentiate products based on the required level of transparency, control and documentation support determined by the nee...

One Quality Policy [VIDEO]

Sigma-Aldrich maintains one quality policy on a global level to provide consistency in vision, maintained across all manufacturing and distribution sites so that the company on the whole is adhering ...

Quality Management Systems [VIDEO]

See how we combine the use of various Quality Management Systems to manage manufacturing and supply chain for our products, as well as ensure data integrity for information provided to customers via ...

Risk Management [VIDEO]

Our Quality Policy means that we are able to provide certain information to end customers based on a product’s Enhanced Quality level, and that information is limited to what we are able to guarantee...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?