EMAIL THIS PAGE TO A FRIEND
G6011 Sigma-Aldrich

Gly-Pro-Gly-Gly

≥97% (HPLC)

  • CAS Number 13054-03-0

  • Empirical Formula (Hill Notation) C11H18N4O5

  • Molecular Weight 286.28

  •  MDL number MFCD00133379

  •  PubChem Substance ID 24895240

Purchase

Description

Biochem/physiol Actions

Inhibitor of dipeptidyl peptidase IV. Inhibits entry of HIV-1 or HIV-2 into T lymphoblastoid and monocytoid cell lines.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?