• USA Home
  • G6887 - Gly-Gly-Arg acetate salt

EMAIL THIS PAGE TO A FRIEND
G6887 Sigma-Aldrich

Gly-Gly-Arg acetate salt

≥97% (HPLC)

  • CAS Number 55033-48-2

  • Empirical Formula (Hill Notation) C10H20N6O4

  • Molecular Weight 288.30

  •  PubChem Substance ID 329799955

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
SMILES string   CC(O)=O.NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(O)=O
InChI   1S/C10H20N6O4.C2H4O2/c11-4-7(17)16-6(2-1-3-14-10(12)13)9(20)15-5-8(18)19;1-2(3)4/h6H,1-5,11H2,(H,15,20)(H,16,17)(H,18,19)(H4,12,13,14);1H3,(H,3,4)/t6-;/m0./s1
InChI key   JBEBGKUWGHTBHG-RGMNGODLSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?