• USA Home
  • G7125 - D-Glucose 6-phosphate barium salt hydrate

EMAIL THIS PAGE TO A FRIEND
G7125 Sigma-Aldrich

D-Glucose 6-phosphate barium salt hydrate

Sigma Grade, 98-100%

Synonym: D(+)-Glucopyranose 6-phosphate barium salt, G-6-P, Robison ester

  • CAS Number 150400-00-3

  • Empirical Formula (Hill Notation) C6H11BaO9P · xH2O

  • Molecular Weight 395.45 (anhydrous basis)

  •  Beilstein/REAXYS Number 5199009

  •  MDL number MFCD00221522

  •  PubChem Substance ID 24895289

Purchase

Properties

grade   Sigma Grade
assay   98-100%
storage temp.   −20°C
SMILES string   [Ba].OC(COP(O)(O)=O)C(O)C(O)C(O)C=O
InChI   1S/C6H13O9P.Ba.2H/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;;;/h1,3-6,8-11H,2H2,(H2,12,13,14);;;
InChI key   XZVFATVUCZXZGZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?