EMAIL THIS PAGE TO A FRIEND
G7897 Sigma-Aldrich

Granuliberin R

≥97% (HPLC)

Synonym: Phe-Gly-Phe-Leu-Pro-Ile-Tyr-Arg-Arg-Pro-Ala-Ser-NH2

  • CAS Number 64704-41-2

  • Empirical Formula (Hill Notation) C69H103N19O14

  • Molecular Weight 1422.68

  •  MDL number MFCD00080191

  •  PubChem Substance ID 24895328

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   IQNSWGKGZHMSIZ-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Mast cell degranulating peptide found in the skin of the frog, Rana rugosa.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?