• USA Home
  • G7920 - β-D-Glucose 1-phosphate bis(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
G7920 Sigma-Aldrich

β-D-Glucose 1-phosphate bis(cyclohexylammonium) salt

  • CAS Number 14034-70-9

  • Empirical Formula (Hill Notation) C6H13O9P · 2C6H13N

  • Molecular Weight 458.48

  •  MDL number MFCD00069780

  •  PubChem Substance ID 329799970

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Glycolysis,
InChI Key   YBPDNWXTAIIEAM-LCFXSEKVSA-N
impurities   α-D-Glucose 1-phosphate, essentially free
storage temp.   −20°C

Description

Biochem/physiol Actions

β-D-Glucose 1-phosphate is a substrate for β-phosphoglucomutase, which converts it to β-glucose 6-phosphate by forming β-glucose 1,6-diphosphate as an intermediate. β-Phosphoglucomutase is utilized in both the maltose and trehalose catabolic pathways of lactic acid bacteria, generating β-D-glucose 1-phosphate as a metabolite.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?