• USA Home
  • G8012 - Guanosine 5′-diphosphate, periodate oxidized sodium salt

EMAIL THIS PAGE TO A FRIEND
G8012 Sigma-Aldrich

Guanosine 5′-diphosphate, periodate oxidized sodium salt

90-95%

Synonym: Guanosine 5′-diphosphate-2′,3′-dialdehyde

Purchase

Properties

assay   90-95%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?