• USA Home
  • G9773 - Gly-Phe-Asp-Leu-Asn-Gly-Gly-Gly-Val-Gly

EMAIL THIS PAGE TO A FRIEND
G9773 Sigma-Aldrich

Gly-Phe-Asp-Leu-Asn-Gly-Gly-Gly-Val-Gly

≥97% (HPLC)

Synonym: Speract

  • CAS Number 76901-59-2

  • Empirical Formula (Hill Notation) C38H57N11O14

  • Molecular Weight 891.92

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Stimulates the respiration, motility, and cyclic nucleotide metabolism of sea urchin spermatozoa

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?