• USA Home
  • H0127 - Hydroxylamine sulfate

EMAIL THIS PAGE TO A FRIEND
H0127 Sigma-Aldrich

Hydroxylamine sulfate

≥99% (titration)

Synonym: Hydroxylammonium sulfate

Purchase

Properties

assay   ≥99% (titration)
mp   170 °C (dec.) (lit.)
SMILES string   NO.NO.OS(O)(=O)=O
InChI   1S/2H3NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H2;(H2,1,2,3,4)
InChI key   VRXOQUOGDYKXFA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2865 8 / PGIII
WGK Germany 
3
RTECS 
NC5425000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?