• USA Home
  • H0648 - 5-Hydroxytryptamine-β-14C creatinine sulfate complex

EMAIL THIS PAGE TO A FRIEND
H0648 Sigma-Aldrich

5-Hydroxytryptamine-β-14C creatinine sulfate complex

~95% (Radiochemical Purity), aqueous ethanol solution

Synonym: Serotonin creatinine sulfate complex

Purchase

Properties

assay   ~95% (Radiochemical Purity)
form   aqueous ethanol solution
extent of labeling   20-60 mCi per mmol
mass shift   M+2
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

serum bottle

Physical form

Aqueous solution containing 2% ethanol

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?