EMAIL THIS PAGE TO A FRIEND
H0762 Sigma-Aldrich

DL-Homophenylalanine

Synonym: (±)-2-Amino-4-phenylbutyric acid

  • CAS Number 1012-05-1

  • Linear Formula C6H5(CH2CH2)CH(NH2)CO2H

  • Molecular Weight 179.22

  •  MDL number MFCD00063092

Purchase

Properties

mp   282 °C (dec.) (lit.)
SMILES string   NC(CCc1ccccc1)C(O)=O
InChI   1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)
InChI key   JTTHKOPSMAVJFE-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

294357 DL-Homophenylalanine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?