EMAIL THIS PAGE TO A FRIEND
H0882 Sigma-Aldrich

4(3H)-Pyrimidinone

Synonym: 4(3H)-Pyrimidone, 4-Hydroxypyrimidine, 4-Pyrimidinol

  • CAS Number 4562-27-0

  • Empirical Formula (Hill Notation) C4H4N2O

  • Molecular Weight 96.09

  •  Beilstein/REAXYS Number 609975

  •  EC Number 224-932-4

  •  MDL number MFCD00167178

Purchase

Properties

InChI Key   DNCYBUMDUBHIJZ-UHFFFAOYSA-N
mp   166-169 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?