• USA Home
  • H1877 - 4-Imidazolemethanol hydrochloride

EMAIL THIS PAGE TO A FRIEND
H1877 Sigma-Aldrich

4-Imidazolemethanol hydrochloride

crystalline

Synonym: 4-(Hydroxymethyl)imidazole hydrochloride

  • CAS Number 32673-41-9

  • Empirical Formula (Hill Notation) C4H6N2O · HCl

  • Molecular Weight 134.56

  •  Beilstein/REAXYS Number 3562041

  •  EC Number 251-150-0

  •  MDL number MFCD00012697

  •  PubChem Substance ID 24278462

Purchase

Properties

form   crystalline
color   off-white to yellow
mp   108-111 °C (lit.)
storage temp.   −20°C
SMILES string   Cl[H].OCc1c[nH]cn1
InChI   1S/C4H6N2O.ClH/c7-2-4-1-5-3-6-4;/h1,3,7H,2H2,(H,5,6);1H
InChI key   WFNASTYGEKUMIY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

219908 4-Imidazolemethanol hydrochloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?