EMAIL THIS PAGE TO A FRIEND
H1880 Sigma-Aldrich

5-Hydroxytryptophol

~98%

Synonym: 2-(5-Hydroxy-3-indolyl)ethyl alcohol

Purchase

Properties

assay   ~98%
storage temp.   −20°C
SMILES string   OCCc1c[nH]c2ccc(O)cc12

Description

Biochem/physiol Actions

Serotonin metabolite

Packaging

Sealed ampule

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
NL8512500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?