• USA Home
  • H2391 - Hydroxylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
H2391 Sigma-Aldrich

Hydroxylamine hydrochloride

ACS reagent, ≥96.0%

Synonym: Hydroxylammonium chloride

  • CAS Number 5470-11-1

  • Linear Formula NH2OH · HCl

  • Molecular Weight 69.49

  •  Beilstein/REAXYS Number 3539763

  •  EC Number 226-798-2

  •  MDL number MFCD00051089

Purchase

Properties

grade   ACS reagent
assay   ≥96.0%
impurities   ≤0.25 meq/g Titratable free acid
ign. residue   ≤0.05%
mp   155-157 °C (dec.) (lit.)
solubility   ethanol: clarity passes test
density   1.67 g/mL at 25 °C (lit.)
anion traces   sulfur compounds (as SO42-): ≤0.005%
cation traces   Fe: ≤5 ppm
  NH4+:, passes test
  heavy metals (as Pb): ≤5 ppm
SMILES string   Cl.NO
InChI   1S/ClH.H3NO/c;1-2/h1H;2H,1H2
InChI key   WTDHULULXKLSOZ-UHFFFAOYSA-N

Description

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Safety & Documentation

Safety Information

Signal word 
Warning
RIDADR 
UN2923 - class 8 - PG 3 - EHS - acidic - Corrosive solids, toxic, n.o.s., HI: all
WGK Germany 
3
RTECS 
NC3675000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?