• USA Home
  • H3380 - Hexamethylphosphoramide

EMAIL THIS PAGE TO A FRIEND
H3380 Sigma-Aldrich

Hexamethylphosphoramide

Synonym: HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide

  • CAS Number 680-31-9

  • Linear Formula [(CH3)2N]3PO

  • Molecular Weight 179.20

  •  Beilstein/REAXYS Number 1099903

  •  EC Number 211-653-8

  •  MDL number MFCD00008303

Purchase

Properties

vapor density   6.18 (vs air)
vapor pressure   0.07 mmHg ( 25 °C)
InChI Key   GNOIPBMMFNIUFM-UHFFFAOYSA-N
refractive index   n20/D 1.459 (lit.)
bp   230-232 °C/740 mmHg(lit.)
mp   7 °C (lit.)
density   1.03 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
TD0875000
Flash Point(F) 
291.2 °F
Flash Point(C) 
144 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H11602 Hexamethylphosphoramide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?