• USA Home
  • H3641 - 2-(n-Hexadecanoylamino)-4-nitrophenol

EMAIL THIS PAGE TO A FRIEND
H3641 Sigma-Aldrich

2-(n-Hexadecanoylamino)-4-nitrophenol

~99% (TLC)

Synonym: 2′-Hydroxy-5′-nitrohexadecananilide

  • CAS Number 60301-87-3

  • Empirical Formula (Hill Notation) C22H36N2O4

  • Molecular Weight 392.53

  •  MDL number MFCD00056113

  •  PubChem Substance ID 24895584

Purchase

Properties

Related Categories Biochemicals and Reagents, Chromogen, Enzyme Substrate, Fatty Acyls, Lipids More...
InChI Key   OXIUBIYNTOOVQI-UHFFFAOYSA-N
assay   ~99% (TLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Webinar - Hydrogels
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?