EMAIL THIS PAGE TO A FRIEND
H4030 Sigma-Aldrich

6-Hydroxyflavanone

  • CAS Number 4250-77-5

  • Empirical Formula (Hill Notation) C15H12O3

  • Molecular Weight 240.25

  •  MDL number MFCD00017485

Purchase

Properties

mp   220-222 °C (dec.) (lit.)
SMILES string   Oc1ccc2OC(CC(=O)c2c1)c3ccccc3
InChI   1S/C15H12O3/c16-11-6-7-14-12(8-11)13(17)9-15(18-14)10-4-2-1-3-5-10/h1-8,15-16H,9H2
InChI key   XYHWPQUEOOBIOW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

419796 6-Hydroxyflavanone, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?