• USA Home
  • H5128 - 8-Hydroxyadenosine 3′:5′-cyclic monophosphate

EMAIL THIS PAGE TO A FRIEND
H5128 Sigma-Aldrich

8-Hydroxyadenosine 3′:5′-cyclic monophosphate

Synonym: 7,8-Dihydro-8-oxoadenosine 3′:5′-cyclic monophosphate, 8-Hydroxy-cyclic AMP

  • CAS Number 31356-95-3

  • Empirical Formula (Hill Notation) C10H12N5O7P

  • Molecular Weight 345.21

  •  MDL number MFCD00079352

  •  PubChem Substance ID 24895671

Purchase

Properties

storage temp.   −20°C
SMILES string   Nc1ncnc2N(C3OC4COP(O)(=O)OC4C3O)C(=O)Nc12
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?