• USA Home
  • H6021 - DL-α-Hydroxybehenic acid

EMAIL THIS PAGE TO A FRIEND
H6021 Sigma-Aldrich

DL-α-Hydroxybehenic acid

≥98% (capillary GC)

Synonym: 2-Hydroxydocosanoic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Fatty Acids and conjugates, Fatty Acyls, Hydroxy Fatty Acids,
InChI Key   RPGJJWLCCOPDAZ-UHFFFAOYSA-N
assay   ≥98% (capillary GC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?