• USA Home
  • H6152 - cis-3-Hydroxy-DL-proline

EMAIL THIS PAGE TO A FRIEND
H6152 Sigma

cis-3-Hydroxy-DL-proline

Synonym: (±)-cis-3-Hydroxypyrrolidine-2-carboxylic acid

  • CAS Number 4298-05-9

  • Empirical Formula (Hill Notation) C5H9NO3

  • Molecular Weight 131.13

  •  Beilstein Registry Number 4130229

  •  MDL number MFCD00143991

Purchase

Properties

InChI Key   BJBUEDPLEOHJGE-DMTCNVIQSA-N
mp   232 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

420174 cis-3-Hydroxy-DL-proline, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?