EMAIL THIS PAGE TO A FRIEND
H6514 Sigma-Aldrich

Heptacosanoic acid

~99% (capillary GC)

Synonym: C27:0, Carboceric acid

  • CAS Number 7138-40-1

  • Linear Formula CH3(CH2)25COOH

  • Molecular Weight 410.72

  •  Beilstein/REAXYS Number 1800778

  •  EC Number 230-436-9

  •  MDL number MFCD00014029

  •  PubChem Substance ID 24895747

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Lipids, Straight Chain Fatty Acids More...
InChI Key   VXZBFBRLRNDJCS-UHFFFAOYSA-N
assay   ~99% (capillary GC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?