• USA Home
  • H6892 - 1-(4-Hydroxybenzyl)imidazole-2-thiol

EMAIL THIS PAGE TO A FRIEND
H6892 Sigma-Aldrich

1-(4-Hydroxybenzyl)imidazole-2-thiol

  • CAS Number 95333-64-5

  • Empirical Formula (Hill Notation) C10H10N2OS

  • Molecular Weight 206.26

  •  MDL number MFCD00079360

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?