• USA Home
  • H6895 - Hydroxylamine phosphate salt (3:1)

EMAIL THIS PAGE TO A FRIEND
H6895 SAFC

Hydroxylamine phosphate salt (3:1)

~97% (titration)

Purchase

Properties

assay   ~97% (titration)
mp   169-171 °C (lit.)
SMILES string   NO.NO.NO.OP(O)(O)=O
InChI   1S/3H3NO.H3O4P/c3*1-2;1-5(2,3)4/h3*2H,1H2;(H3,1,2,3,4)
InChI key   XBUFCZMOAHHGMX-UHFFFAOYSA-N

Description

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3262 - class 8 - PG 2 - EHS - Corrosive solid, basic, inorganic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?