• USA Home
  • H7524 - Methyl (±)-2-hydroxytetracosanoate

EMAIL THIS PAGE TO A FRIEND
H7524 Sigma-Aldrich

Methyl (±)-2-hydroxytetracosanoate

>98% (GC)

Synonym: DL-2-Hydroxytetracosanoic acid methyl ester

  • CAS Number 2433-95-6

  • Empirical Formula (Hill Notation) C25H50O3

  • Molecular Weight 398.66

  •  MDL number MFCD00214302

  •  PubChem Substance ID 24895797

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acyls, Fatty Esters, Hydroxy fatty acids, Lipids,
assay   >98% (GC)
form   solid
functional group   ester
shipped in   ambient
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCCCCCC(O)C(=O)OC
InChI   1S/C25H50O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(26)25(27)28-2/h24,26H,3-23H2,1-2H3
InChI key   MSVLAXSCPVNLFM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?