• USA Home
  • H7769 - gp120 Fragment 254-274

EMAIL THIS PAGE TO A FRIEND
H7769 Sigma-Aldrich

gp120 Fragment 254-274

≥95% (HPLC)

Purchase

Description

Amino Acid Sequence

Cys-Thr-His-Gly-Ile-Arg-Pro-Val-Val-Ser-Thr-Gln-Leu-Leu-Leu-Asn-Gly-Ser-Leu-Ala-Glu

General description

The gp120 (glycoprotein-120) is a HIV (human immunodeficiency virus)-1 envelope glycoprotein. The entry of the virus in cells requires association between gp120 and cell surface receptors, CD4 (cluster of differentiation 4) and chemokine receptor. The gp120 fragment (254-274) is conserved in all strains of HIV.

Biochem/physiol Actions

Fragment from the second conserved domain of gp120. Important for HIV infectivity and antibody neutralization.

Packaging

Packaged under argon

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?