EMAIL THIS PAGE TO A FRIEND
H8034 Sigma

HE-NECA

solid, ≥98% (HPLC)

Synonym: 2-(1-Hexyn-1-yl)adenosine-5′-N-ethyluronamide, 2-Hexynyl-5′-ethylcarboxamidoadenosine

Purchase

Properties

Related Categories Adenosines/P2 Nucleotide Receptors (Purinergics), Agonists, Cell Biology, Cell Signaling and Neuroscience, Neuroscience,
InChI Key   FDEACFAXFCKCHZ-MOROJQBDSA-N
assay   ≥98% (HPLC)
form   solid
color   white
solubility   DMSO: >10 mg/mL
  H2O: insoluble
storage temp.   2-8°C
Gene Information   human ... ADORA2A(135), ADORA2B(136)

Description

Biochem/physiol Actions

A2 adenosine receptor agonist

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C141 CGS-21680 hydrochloride hydrate, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?