• USA Home
  • H8260 - S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine

EMAIL THIS PAGE TO A FRIEND
H8260 Sigma-Aldrich

S-(2-Hydroxy-5-nitrobenzyl)-6-thioinosine

Synonym: 6-[(2-Hydroxy-5-nitrobenzyl)thio]-9-β-D-ribofuranosylpurine

  • CAS Number 56964-73-9

  • Empirical Formula (Hill Notation) C17H17N5O7S

  • Molecular Weight 435.41

  •  MDL number MFCD00005744

  •  PubChem Substance ID 24895821

Purchase

Properties

form   solid
storage temp.   −20°C
SMILES string   OC[C@@H]1O[C@@H]([C@@H](O)[C@H]1O)n2cnc3c(SCc4cc(ccc4O)[N+]([O-])=O)ncnc23
InChI   1S/C17H17N5O7S/c23-4-11-13(25)14(26)17(29-11)21-7-20-12-15(21)18-6-19-16(12)30-5-8-3-9(22(27)28)1-2-10(8)24/h1-3,6-7,11,13-14,17,23-26H,4-5H2/t11-,13-,14-,17-/m1/s1
InChI key   UNTSTPIAQOSGHD-LSCFUAHRSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?