• USA Home
  • H9391 - 3-Hydroxyanthranilic acid

EMAIL THIS PAGE TO A FRIEND
H9391 Sigma-Aldrich

3-Hydroxyanthranilic acid

Synonym: 2-Amino-3-hydroxybenzoic acid

Purchase

Properties

application(s)   cell culture | mammalian: suitable
  peptide synthesis: suitable
mp   240 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   Nc1c(O)cccc1C(O)=O
InChI   1S/C7H7NO3/c8-6-4(7(10)11)2-1-3-5(6)9/h1-3,9H,8H2,(H,10,11)
InChI key   WJXSWCUQABXPFS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG2625000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

148776 3-Hydroxyanthranilic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?