• USA Home
  • H9396 - 5(S)-Hydroperoxy-(6E,8Z,11Z,14Z)-eicosatetraenoic acid

EMAIL THIS PAGE TO A FRIEND
H9396 Sigma-Aldrich

5(S)-Hydroperoxy-(6E,8Z,11Z,14Z)-eicosatetraenoic acid

~100 μg/mL in ethanol, ~98%

Synonym: 5(S)-HPETE

  • CAS Number 71774-08-8

  • Empirical Formula (Hill Notation) C20H32O4

  • Molecular Weight 336.47

  •  MDL number MFCD00036929

Purchase

Properties

assay   ~98%
concentration   ~100 μg/mL in ethanol
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
3
RTECS 
JX3867000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?