• USA Home
  • I1765 - Indanomycin from Streptomyces antibioticus

EMAIL THIS PAGE TO A FRIEND
I1765 Sigma-Aldrich

Indanomycin from Streptomyces antibioticus

~98% (TLC)

Synonym: X-14547A

  • CAS Number 66513-28-8

  • Empirical Formula (Hill Notation) C31H43NO4

  • Molecular Weight 493.68

  •  PubChem Substance ID 329815357

Purchase

Properties

InChI Key   BAIPOTOKPGDCHA-MWBHXQFBSA-N
assay   ~98% (TLC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
UP6100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?