EMAIL THIS PAGE TO A FRIEND
I2503 Sigma

DL-Isoserine

Synonym: (±)-3-Amino-2-hydroxypropionic acid, DL-2-Hydroxy-3-aminopropionic acid

  • CAS Number 632-12-2

  • Linear Formula H2NCH2CH(OH)CO2H

  • Molecular Weight 105.09

  •  Beilstein Registry Number 1721413

  •  MDL number MFCD00008138

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

286338 DL-Isoserine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?