• USA Home
  • I4002 - 2′,3′-O-Isopropylideneadenosine

EMAIL THIS PAGE TO A FRIEND
I4002 Sigma

2′,3′-O-Isopropylideneadenosine

≥99% (HPLC)

  • CAS Number 362-75-4

  • Empirical Formula (Hill Notation) C13H17N5O4

  • Molecular Weight 307.31

  •  Beilstein Registry Number 43435

  •  EC Number 206-650-3

  •  MDL number MFCD00005756

Purchase

Properties

InChI Key   LCCLUOXEZAHUNS-WOUKDFQISA-N
assay   ≥99% (HPLC)
mp   221-222 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

I22404 2′,3′-O-Isopropylideneadenosine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?