EMAIL THIS PAGE TO A FRIEND
I4897 Supelco

Lithium ionophore II

Synonym: N,N,N′,N′-Tetraisobutyl-cis-cyclohexane-1,2-dicarboxamide, ETH 1644

  • CAS Number 80547-18-8

  • Empirical Formula (Hill Notation) C24H46N2O2

  • Molecular Weight 394.63

  •  Beilstein/REAXYS Number 6428727

  •  MDL number MFCD00042968

  •  PubChem Substance ID 24896064

Purchase

Properties

storage temp.   2-8°C
SMILES string   CC(C)CN(CC(C)C)C(=O)[C@H]1CCCC[C@H]1C(=O)N(CC(C)C)CC(C)C
InChI   1S/C24H46N2O2/c1-17(2)13-25(14-18(3)4)23(27)21-11-9-10-12-22(21)24(28)26(15-19(5)6)16-20(7)8/h17-22H,9-16H2,1-8H3/t21-,22+
InChI key   GPFAWYFTCKIPRA-SZPZYZBQSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?