• USA Home
  • I5514 - D-myo-Inositol 1,4,5,6-tetrakis(phosphate) potassium salt

EMAIL THIS PAGE TO A FRIEND
I5514 Sigma-Aldrich

D-myo-Inositol 1,4,5,6-tetrakis(phosphate) potassium salt

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Monosaccharide More...
Quality Level   100
storage temp.   −20°C
SMILES string   [K+].[K+].[K+].[K+].[K+].[K+].[K+].[K+].O[C@@H]1[C@H](O)[C@@H](OP([O-])([O-])=O)[C@H](OP([O-])([O-])=O)[C@@H](OP([O-])([O-])=O)[C@@H]1OP([O-])([O-])=O
InChI   1S/C6H16O18P4.8K/c7-1-2(8)4(22-26(12,13)14)6(24-28(18,19)20)5(23-27(15,16)17)3(1)21-25(9,10)11;;;;;;;;/h1-8H,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20);;;;;;;;/q;8*+1/p-8/t1-,2+,3-,4-,5+,6+;;;;;;;;/m1......../s1
InChI key   KOARMWMDZTZZSB-FPQWBLQBSA-F

Description

Biochem/physiol Actions

Component of lipid signaling pathway; shown to antagonize the negative regulation of calcium-mediated chloride secretion by epidermal growth factor; also shown to bind to pleckstrin homology (PH) domain of p130

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?